Anti-RENBP Rabbit Polyclonal Antibody

Lieferant: ProSci Inc.
13-373
PRSI13-373EA 419 EUR
PRSI13-373
Anti-RENBP Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: RENBP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR