Anti-RPRM Rabbit Polyclonal Antibody

Lieferant: ProSci Inc.
13-374
PRSI13-374EA 419 EUR
PRSI13-374
Anti-RPRM Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: RPRM
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR