Anti-SLC22A4 Rabbit Polyclonal Antibody

Lieferant: ProSci Inc.
13-685
PRSI13-685EA 419 EUR
PRSI13-685
Anti-SLC22A4 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SLC22A4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR