Anti-SLC27A2 Rabbit Polyclonal Antibody

Lieferant: ProSci Inc.
13-793
PRSI13-793EA 419 EUR
PRSI13-793
Anti-SLC27A2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SLC27A2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR