Anti-U2AF1 Rabbit Polyclonal Antibody

Lieferant: ProSci Inc.
14-831
PRSI14-831EA 419 EUR
PRSI14-831
Anti-U2AF1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: U2AF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR