Anti-GPD1L Rabbit Polyclonal Antibody

Lieferant: ProSci Inc.
15-518
PRSI15-518EA 419 EUR
PRSI15-518
Anti-GPD1L Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: GPD1L
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR