Anti-CCDC83 Rabbit Polyclonal Antibody

Lieferant: ProSci Inc.
15-554
PRSI15-554EA 419 EUR
PRSI15-554
Anti-CCDC83 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CCDC83
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR