Anti-TRH Rabbit Polyclonal Antibody

Lieferant: ProSci Inc.
15-561
PRSI15-561EA 419 EUR
PRSI15-561
Anti-TRH Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: TRH
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR