Anti-TPD52L3 Rabbit Polyclonal Antibody

Lieferant: ProSci Inc.
16-100
PRSI16-100EA 419 EUR
PRSI16-100
Anti-TPD52L3 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: TPD52L3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR