Anti-DPP9 Rabbit Polyclonal Antibody

Lieferant: ProSci Inc.
16-102
PRSI16-102EA 419 EUR
PRSI16-102
Anti-DPP9 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: DPP9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR