Anti-PEX12 Rabbit Polyclonal Antibody

Lieferant: ProSci Inc.
16-406
PRSI16-406EA 419 EUR
PRSI16-406
Anti-PEX12 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PEX12
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR