Anti-U2AF2 Rabbit Polyclonal Antibody

Lieferant: ProSci Inc.
18-399
PRSI18-399EA 419 EUR
PRSI18-399
Anti-U2AF2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: U2AF2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR