Anti-SNCG Rabbit Polyclonal Antibody

Lieferant: ProSci Inc.
18-688
PRSI18-688EA 419 EUR
PRSI18-688
Anti-SNCG Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SNCG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR