Anti-ORF3a Sheep Polyclonal Antibody

Lieferant: ProSci Inc.
20-281
PRSI20-281EA 747 EUR
PRSI20-281
Anti-ORF3a Sheep Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ORF3a
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype:
Reactivity: Virus
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR