Anti-SPR Rabbit Polyclonal Antibody

Lieferant: ProSci Inc.
23-285
PRSI23-285EA 419 EUR
PRSI23-285
Anti-SPR Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SPR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Monkey
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR