Anti-PSA Recombinant Antibody [Clone: 735]

Lieferant: ProSci Inc.
24-268
PRSI24-268EA 737 EUR
PRSI24-268
Anti-PSA Recombinant Antibody [Clone: 735]
Antikörper
Type: Primary
Antigen: PSA
Clonality: Recombinant
Clone: 735
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG2a kappa
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR