Anti-PSA Recombinant Antibody [Clone: 735]

Lieferant: ProSci Inc.
24-269
PRSI24-269EA 927 EUR
PRSI24-269
Anti-PSA Recombinant Antibody [Clone: 735]
Antikörper
Type: Primary
Antigen: PSA
Clonality: Recombinant
Clone: 735
Conjugation: Unconjugated
Host: Recombinant
Isotype: IgG kappa
Reactivity: Human;Mouse;Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR