Anti-Ms4a1 Recombinant Antibody [Clone: 18B12]

Lieferant: ProSci Inc.
24-638
PRSI24-638EA 737 EUR
PRSI24-638
Anti-Ms4a1 Recombinant Antibody [Clone: 18B12]
Antikörper
Type: Primary
Antigen: membrane-spanning 4-domains, subfamily A, member 1
Clonality: Recombinant
Clone: 18B12
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG kappa
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR