Anti-OTX1 Rabbit Polyclonal Antibody

Lieferant: ProSci Inc.
25-475
PRSI25-475EA 660 EUR
PRSI25-475
Anti-OTX1 Rabbit Polyclonal Antibody
Antikörper
OTX1 probably plays a role in the development of the brain and the sense organs. OTX1 can bind to the BCD target sequence (BTS): 5'-TCTAATCCC-3'.This gene encodes a member of the bicoid sub-family of homeodomain-containing transcription factors. The encoded protein acts as a transcription factor and may play a role in brain and sensory organ development. A similar protein in mice is required for proper brain and sensory organ development and can cause epilepsy.

Antibody is purified by peptide affinity chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: OTX1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Dog, Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR