Anti-ZNF414 Rabbit Polyclonal Antibody

Lieferant: ProSci Inc.
25-546
PRSI25-546EA 660 EUR
PRSI25-546
Anti-ZNF414 Rabbit Polyclonal Antibody
Antikörper
ZNF414 may be involved in transcriptional regulation.

Antibody is purified by peptide affinity chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: ZNF414
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR