Anti-EVX2 Rabbit Polyclonal Antibody

Lieferant: ProSci Inc.
25-584
PRSI25-584EA 660 EUR
PRSI25-584
Anti-EVX2 Rabbit Polyclonal Antibody
Antikörper
EVX2 belongs to the even-skipped homeobox family. It contains 1 homeobox DNA-binding domain.117-kb microdeletion removing HOXD9-HOXD13 and EVX2 causes synpolydactyly.

Antibody is purified by peptide affinity chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: EVX2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR