Anti-STS Rabbit Polyclonal Antibody

Lieferant: ProSci Inc.
25-714
PRSI25-714EA 660 EUR
PRSI25-714
Anti-STS Rabbit Polyclonal Antibody
Antikörper
STS catalyzes the conversion of sulfated steroid precursors to estrogens during pregnancy. The protein is found in the endoplasmic reticulum, where it acts as a homodimer. Mutations in its gene are known to cause X-linked ichthyosis The protein encoded by this gene catalyzes the conversion of sulfated steroid precursors to estrogens during pregnancy. The encoded protein is found in the endoplasmic reticulum, where it acts as a homodimer. Mutations in this gene are known to cause X-linked ichthyosis (XLI). Publication Note: This RefSeq record includes a subset of the publications that are available for this gene. Please see the Entrez Gene record to access additional publications.

Antibody is purified by peptide affinity chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: STS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR