Anti-GGCX Rabbit Polyclonal Antibody

Lieferant: ProSci Inc.
25-950
PRSI25-950EA 660 EUR
PRSI25-950
Anti-GGCX Rabbit Polyclonal Antibody
Antikörper
GGCX is an enzyme which catalyzes the posttranslational modification of vitamin K-dependent protein. Many of these vitamin K-dependent proteins are involved in coagulation so the function of the encoded enzyme is essential for hemostasis. Mutations in this gene are associated with vitamin K-dependent coagulation defect and PXE-like disorder with multiple coagulation factor deficiency.Gamma-glutamyl carboxylase accomplishes the posttranslational modification required for the activity of all of the vitamin K-dependent proteins (Wu et al., 1991 [PubMed 1749935]). These include some of the blood coagulation and anticoagulation proteins as well as bone gamma-carboxyglutamic acid (Gla) protein (BGLAP; MIM 112260) and bone matrix protein (MGP; MIM 154870).

Antibody is purified by peptide affinity chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: GGCX
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR