Anti-NAAA Rabbit Polyclonal Antibody

Lieferant: ProSci Inc.
26-026
PRSI26-026EA 660 EUR
PRSI26-026
Anti-NAAA Rabbit Polyclonal Antibody
Antikörper
ASAHL is an N-acylethanolamine-hydrolyzing enzyme which is highly similar to acid ceramidase. This gene encodes an N-acylethanolamine-hydrolyzing enzyme which is highly similar to acid ceramidase. Multiple transcript variants encoding different isoforms have been found for this gene.

Antibody is purified by peptide affinity chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: NAAA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR