Anti-TMTC4 Rabbit Polyclonal Antibody

Lieferant: ProSci Inc.
26-029
PRSI26-029EA 660 EUR
PRSI26-029
Anti-TMTC4 Rabbit Polyclonal Antibody
Antikörper
It belongs to the TMTC family. Its exact function remains unknown.

Antibody is purified by peptide affinity chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: TMTC4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR