Anti-CEACAM4 Rabbit Polyclonal Antibody

Lieferant: ProSci Inc.
26-036
PRSI26-036EA 660 EUR
PRSI26-036
Anti-CEACAM4 Rabbit Polyclonal Antibody
Antikörper
CEACAM4 belongs to the immunoglobulin superfamily.

Antibody is purified by peptide affinity chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: CEACAM4
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR