Anti-GALNT10 Rabbit Polyclonal Antibody

Lieferant: ProSci Inc.
26-349
PRSI26-349EA 660 EUR
PRSI26-349
Anti-GALNT10 Rabbit Polyclonal Antibody
Antikörper
GALNT10 belongs to the polypeptide N-acetylgalactosaminyltransferase (pp-GalNAc-T) family. Polypeptide GalNAc transferases initiate the synthesis of mucin-type oligosaccharides by transferring GalNAc from UDP-GalNAc to the hydroxyl group of either a serine or threonine residue on the polypeptide acceptor. Following expression in insect cells, recombinant GalNAc transferase 10 showed significant GalNAcT activity toward mucin-derived peptides, and it utilized both nonglycosylated and glycosylated peptide substrates.This gene belongs to the polypeptide N-acetylgalactosaminyltransferase (pp-GalNAc-T) gene family. Polypeptide GalNAc transferases initiate the synthesis of mucin-type oligosaccharides by transferring GalNAc from UDP-GalNAc to the hydroxyl group of either a serine or threonine residue on the polypeptide acceptor. Following expression in insect cells, recombinant GalNAc transferase 10 showed significant GalNAcT activity toward mucin-derived peptides, and it utilized both nonglycosylated and glycosylated peptide substrates. Two transcript variants encoding distinct isoforms have been identified for this gene.

Antibody is purified by peptide affinity chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: GALNT10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR