Anti-PIP5KL1 Rabbit Polyclonal Antibody

Lieferant: ProSci Inc.
26-764
PRSI26-764EA 660 EUR
PRSI26-764
Anti-PIP5KL1 Rabbit Polyclonal Antibody
Antikörper
PIP5KL1 may act as a scaffold to localize and regulate type I PI (4)P 5-kinases to specific compartments within the cell, where they generate PI (4,5)P2 for actin nucleation, signaling and scaffold protein recruitment and conversion to PI (3,4,5)P3.

Antibody is purified by peptide affinity chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: PIP5KL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR