Anti-FAAH2 Rabbit Polyclonal Antibody

Lieferant: ProSci Inc.
26-772
PRSI26-772EA 660 EUR
PRSI26-772
Anti-FAAH2 Rabbit Polyclonal Antibody
Antikörper
Fatty acid amide hydrolases, such as FAAH1 and FAAH2, hydrolyze primary fatty acid amide substrates and may play a role in fatty acid catabolism.Fatty acid amide hydrolases, such as FAAH1 (FAAH; MIM 602935) and FAAH2, hydrolyze primary fatty acid amide substrates (e.g., oleamide) and may play a role in fatty acid catabolism (Wei et al., 2006 [PubMed 17015445]).

Antibody is purified by peptide affinity chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: FAAH2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR