Anti-ITPK1 Rabbit Polyclonal Antibody

Lieferant: ProSci Inc.
26-957
PRSI26-957EA 660 EUR
PRSI26-957
Anti-ITPK1 Rabbit Polyclonal Antibody
Antikörper
ITPK1 is the kinase that can phosphorylate various inositol polyphosphate such as Ins (3,4,5,6)P4 or Ins (1,3,4)P3. It may also act as an isomerase that interconverts the inositol tetraphosphate isomers Ins (1,3,4,5)P4 and Ins (1,3,4,6)P4 in the presence of ADP and magnesium. It probably acts as the rate-limiting enzyme of the InsP6 pathway. ITPK1 modifies TNF-alpha-induced apoptosis by interfering with the activation of TNFRSF1A-associated death domain.

Antibody is purified by peptide affinity chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: ITPK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR