Anti-MMACHC Rabbit Polyclonal Antibody

Lieferant: ProSci Inc.
26-997
PRSI26-997EA 660 EUR
PRSI26-997
Anti-MMACHC Rabbit Polyclonal Antibody
Antikörper
The function of C2orf25 remains unknown.Vitamin B12 (cobalamin) is an essential cofactor in several metabolic pathways. Intracellular conversion of cobalamin to adenosylcobalamin in mitochondria and to methylcobalamin in cytoplasm is necessary for homeostasis of methylmalonic acid and homocysteine. C2ORF25 encodes a protein involved in an early step of cobalamin metabolism (Coelho et al., 2008 [PubMed 18385497]).

Antibody is purified by peptide affinity chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: MMACHC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR