Anti-TSPYL4 Rabbit Polyclonal Antibody

Lieferant: ProSci Inc.
27-007
PRSI27-007EA 660 EUR
PRSI27-007
Anti-TSPYL4 Rabbit Polyclonal Antibody
Antikörper
TSPYL4 belongs to the nucleosome assembly protein (NAP) family. The functions of TSPYL4 remain unknown.

Antibody is purified by peptide affinity chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: TSPYL4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR