Anti-HSPA2 Rabbit Polyclonal Antibody
Lieferant: ProSci Inc.
27-009
PRSI27-009EA 660 EUR
PRSI27-009
Anti-HSPA2 Rabbit Polyclonal Antibody
Antikörper
HSPA2 belongs to the heat shock protein 70 family.In cooperation with other chaperones, HSPA2 stabilize preexistent proteins against aggregation and mediate the folding of newly translated polypeptides in the cytosol as well as within organelles. These chaperones participate in all these processes through their ability to recognize nonnative conformations of other proteins. They bind extended peptide segments with a net hydrophobic character exposed by polypeptides during translation and membrane translocation, or following stress-induced damage.
Antibody is purified by peptide affinityMore Product Information