Anti-TFAP2D Rabbit Polyclonal Antibody

Lieferant: ProSci Inc.
27-285
PRSI27-285EA 601 EUR
PRSI27-285
Anti-TFAP2D Rabbit Polyclonal Antibody
Antikörper
TFAP2D has significantly high homology to transcription factor AP-2 gene of human. It is highly expressed in adult thymus, prostate, small intestine, skeletal muscle, placenta, brain, and testis tissues

Antibody is purified by protein A chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 100 μL of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: TFAP2D
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat, Dog, Zebrafish
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR