Anti-FOXP3 Rabbit Polyclonal Antibody

Lieferant: ProSci Inc.
27-688
PRSI27-688EA 601 EUR
PRSI27-688
Anti-FOXP3 Rabbit Polyclonal Antibody
Antikörper
Forkhead box protein P3 (FOXP3, Scurfin, Zinc finger protein JM2) encodes a novel member of the forkhead family of transcription factors. It presumably represses transcription, playing a paramount role in determining the amplitude of the response of CD4 T cells to activation

Antibody is supplied as total IgG.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 200 μL of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: FOXP3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR