Anti-PCYOX1 Rabbit Polyclonal Antibody

Lieferant: ProSci Inc.
27-767
PRSI27-767EA 601 EUR
PRSI27-767
Anti-PCYOX1 Rabbit Polyclonal Antibody
Antikörper
Prenylated proteins contain one of two isoprenoid lipids, either the 15-carbon farnesyl or the 20-carbon geranylgeranyl, covalently attached to cysteine residues at or near their C terminus. PCYOX1 is capable of cleaving the thioether bond of prenylcysteines. The enzyme was isolated from bovine brain membranes and exhibits an apparent molecular mass of 63 kDa. This is a specific enzyme involved in prenylcysteine metabolism in mammalian cells, most likely comprising the final step in the degradation of prenylated proteins.

Antibody is purified by protein A chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 100 μL of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: PCYOX1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR