Anti-MED7 Rabbit Polyclonal Antibody

Lieferant: ProSci Inc.
27-934
PRSI27-934EA 660 EUR
PRSI27-934
Anti-MED7 Rabbit Polyclonal Antibody
Antikörper
CRSP9 is a subunit of the CRSP (cofactor required for SP1 activation) complex, which, along with TFIID, is required for efficient activation by SP1. CRSP9 is also a component of other multisubunit complexes e.g. thyroid hormone receptor- (TR-) associated p

Antibody is purified by peptide affinity chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: MED7
Clonality: Polyclonal
Clone:
Conjugation:More Product Information
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR