Anti-CHAMP1 Rabbit Polyclonal Antibody

Lieferant: ProSci Inc.
27-948
PRSI27-948EA 660 EUR
PRSI27-948
Anti-CHAMP1 Rabbit Polyclonal Antibody
Antikörper
The CD233 gene is located on chromosome 17q21-q22 and is part of the anion exchanger (AE) family. CD233 is expressed in the erythrocyte plasma membrane where it functions as a chloride/bicarbonate exchanger involved in carbon dioxide transport from tissues to lungs. The protein comprises two domains that are structurally and functionally distinct. The N-terminal 40kDa domain is located in the cytoplasm and acts as an attachment site for the red cell skeleton by binding ankyrin. The glycosylated C-terminal membrane-associated domain contains 12-14 membrane spanning segments and carries out the stilbene disulphonate-sensitive exchange transport of anions. The cytoplasmic tail at the extreme C-terminus of the membrane domain binds carbonic anhydrase II. CD233 associates with the red cell membrane protein glycophorin A and this association promotes the correct folding and translocation of CD233. CD233 is predominantly dimeric but forms tetramers in the presence of ankyrin. Many CD233 mutations are known in man and these mutations can lead to two types of disease; destabilization of red cell membrane leading to hereditary spherocytosis, and defective kidney acid secretion leading to distal renal tubular acidosis. Other CD233 mutations that do not give rise to disease result in novel blood group antigens, which form the Diego blood group system. Southeast Asian ovalocytosis (SAO, Melanesian ovalocytosis) results from the heterozygous presence of a deletion in the CD233 protein and is common in areas where Plasmodium falciparum malaria is endemic. One CD233 null human is known also with very severe anemia and nephrocalcinosis [PROW].

Antibody is purified by peptide affinity chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: CHAMP1
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR