ETV5 Contains 1 ETS DNA-binding domain and belongs to the ETS family. The ETV5 gene expression is regulated by the conventional PKC (cPKC) pathway.ETV5 is subject to SUMO modification and this post-translational modification causes inhibition of transcription-enhancing activity Phosphorylated ETV5 and the actin cytoskeleton regulate CD44-mediated hyaluronan binding in myeloid cells. ERMs (ezrin/radixin/moesin) function as adaptor molecules in the interactions of adhesion receptors and intracellular tyrosine kinases. ETV5 can cooperate with c-Jun and has a role in progression of breast cancer.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: ETV5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat