Anti-STS Rabbit Polyclonal Antibody

Lieferant: ProSci Inc.
29-629
PRSI29-629EA 601 EUR
PRSI29-629
Anti-STS Rabbit Polyclonal Antibody
Antikörper
STS catalyzes the conversion of sulfated steroid precursors to estrogens during pregnancy. The protein is found in the endoplasmic reticulum, where it acts as a homodimer. Mutations in its gene are known to cause X-linked ichthyosisThe protein encoded by this gene catalyzes the conversion of sulfated steroid precursors to estrogens during pregnancy. The encoded protein is found in the endoplasmic reticulum, where it acts as a homodimer. Mutations in this gene are known to cause X-linked ichthyosis (XLI).

Antibody is purified by protein A chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 100 μL of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: STS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR