Anti-FAM107A Rabbit Polyclonal Antibody

Lieferant: ProSci Inc.
29-740
PRSI29-740EA 601 EUR
PRSI29-740
Anti-FAM107A Rabbit Polyclonal Antibody
Antikörper
When FAM107A is transfected into cell lines in which it is not expressed, it suppresses cell growth. It may play a role in tumor development.

Antibody is purified by protein A chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 100 μL of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: FAM107A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR