Anti-SLC29A2 Rabbit Polyclonal Antibody

Lieferant: ProSci Inc.
29-910
PRSI29-910EA 601 EUR
PRSI29-910
Anti-SLC29A2 Rabbit Polyclonal Antibody
Antikörper
SLC29A2 mediates equilibrative transport of purine, pyrimidine nucleosides and the purine base hypoxanthine. It is less sensitive than SLC29A1 to inhibition by nitrobenzylthioinosine (NBMPR), dipyridamole, dilazep and draflazine.

Antibody is purified by protein A chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 100 μL of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: SLC29A2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

TECHNISCHE DATEN

Antigen-Symbol SLC29A2
Environmentally Preferable
Antigen-Name Solute Carrier Family 29 Member A2
Antigen-Synonyme Solute carrier family 29 member 2|Equilibrative nucleoside transporter 2|HNP36|ENT2|DER12
Klonalität Polyklonal
Konjugation Unconjugated
Reaktivität Human
Wirt Rabbit
Isotyp
Western blot Yes
ELISA Yes
ImmunoChemie Yes
Immunogen A synthetic peptide corresponding a region of human SLC29A2.
Form Lyophilized

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR