Anti-CBS Rabbit Polyclonal Antibody

Lieferant: ProSci Inc.
30-110
PRSI30-110EA 601 EUR
PRSI30-110
Anti-CBS Rabbit Polyclonal Antibody
Antikörper
CBS is involved in the transsulfuration pathway. The first step of this pathway, from homocysteine to cystathionine, is catalyzed by this protein. CBS deficiency can cause homocystinuria which affects many organs and tissues, including the eyes and the skeletal, vascular and central nervous systems.The protein encoded by this gene is involved in the transsulfuration pathway. The first step of this pathway, from homocysteine to cystathionine, is catalyzed by this protein. CBS deficiency can cause homocystinuria which affects many organs and tissues, including the eyes and the skeletal, vascular and central nervous systems.

Antibody is purified by protein A chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 100 μL of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: CBS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Dog, Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR