Anti-NANOG Rabbit Polyclonal Antibody

Lieferant: ProSci Inc.
30-340
PRSI30-340EA 660 EUR
PRSI30-340
Anti-NANOG Rabbit Polyclonal Antibody
Antikörper
NANOG is a new marker for testicular carcinoma in situ and germ cell tumors. Gene knockdown of Nanog promotes differentiation, thereby demonstrating a role for these factors in human embryonic stem cell self-renewal.

Antibody is purified by peptide affinity chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: NANOG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR