Anti-RABGGTA Rabbit Polyclonal Antibody

Lieferant: ProSci Inc.
30-512
PRSI30-512EA 660 EUR
PRSI30-512
Anti-RABGGTA Rabbit Polyclonal Antibody
Antikörper
RABGGTA belongs to the protein prenyltransferase subunit alpha family and contains 6 PFTA repeats. RABGGTA catalyzes the transfer of a geranyl-geranyl moiety from geranyl-geranyl pyrophosphate to both cysteines in Rab proteins with an -XXCC, -XCXC and -CCXX C-terminal, such as RAB1A, RAB3A and RAB5A respectively.

Antibody is purified by peptide affinity chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: RABGGTA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR