Anti-MCL1 Mouse Monoclonal Antibody

Lieferant: ProSci Inc.
32-191
PRSI32-191EA 509 EUR
PRSI32-191
Anti-MCL1 Mouse Monoclonal Antibody
Antikörper
Mcl-1 (Myeloid cell leukemia-1) is Bcl-2-related and was identified as an early-induction gene that increased in expression during the differentiation of human myeloblastic leukemia cell ML-1, or exposure to different DNA damaging agents. The level of Mcl-1 is decreased in peripheral B lymphocytes undergoing apoptosis following treatment with apoptotic stimuli such as TGF-alpha 1 and forskolin. Expression of Mcl-1 is able to delay apoptosis induced by over-expression of c-myc in CHO 5AHSmyc cells. In hematopoietic FDC-P1 cells, Mcl-1 interacts with another Bcl-2-related protein, Bax, and prolongs cell viability after treatment with different apoptotic reagents.This monoclonal antibody detected a 37kd MCL1 in BCBL-1 cell lysate.

Antibody is purified by protein G affinity chromatography.





Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: MCL1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR