Anti-NTSR2 Rabbit Polyclonal Antibody

Lieferant: ProSci Inc.
33-027
PRSI33-027EA 661 EUR
PRSI33-027
Anti-NTSR2 Rabbit Polyclonal Antibody
Antikörper
Neurotensin is a central nervous system protein involved with the regulation and release of Luteinizing hormone (Lutropin), which regulates ovulation in females and testosterone production in males, and Prolactin (Luteotropin), which enables milk production in female mammals. The GPCR Neurotensin receptor 2 (NTSR2) is one of three receptors for Neurotensin (NTSR1-3). Unlike NTSR1, NTSR2 has only a low affinity for Neurotensin, but high affinity for levocabastine, a histamine H1 receptor antagonist that can block Neurotensin binding to NTSR2 but not affect its binding to NTSR1.

Western blot: 1-3 µg/ml Provided assay concentrations are sµggestions only, Neurotensin Receptor 2 antibody titration may be required for optimal results.

Type: Primary
Antigen: NTSR2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR