Anti-ATG4B Rabbit Polyclonal Antibody

Lieferant: ProSci Inc.
33-064
PRSI33-064EA 661 EUR
PRSI33-064
Anti-ATG4B Rabbit Polyclonal Antibody
Antikörper
Autophagy is the cellular mechanism of degradation and turnover of unnecessary or dysfunctional cellular components. The unneeded components are engulfed in vesicles called autophagosomes, where they are broken down and recycled. The ‘autophagy-related gene’ ATG4B is a regulator of autophagasome maturation. The microtubule-associated protein LC3-I is lipidated (called LC3-II in its lipidated state) and recruited to the autophagosomal membrane where it is of vital importance in the maturation of autophagosomes. ATG4B regulates LC3-II availability by modulating LC3-I lipidation.

Western blot: 4-6 µg/ml Provided assay concentrations are sµggestions only, ATG4B antibody titration may be required for optimal results.

Type: Primary
Antigen: ATG4B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR