Anti-MUC5AC Mouse Monoclonal Antibody [clone: 45M1]

Lieferant: ProSci Inc.
33-176
PRSI33-176EA 773 EUR
PRSI33-176
Anti-MUC5AC Mouse Monoclonal Antibody [clone: 45M1]
Antikörper
This antibody recognises the peptide core of gastric mucin M1 (>1,000 kDa), now identified as Mucin-5AC or MUC5AC. The MUC5AC epitope is destroyed by beta-mercaptoethanol and proteases but not by periodate treatment. MUC5AC antibody reacts with the gastric epithelium of normal human gastrointestinal tract as well as with the precancerous and cancerous colon but not with normal adult colon. It also reacts with fetal colonic mucosa. Resurgence of gastric mucin reactivity during colonic carcinogenesis is due to re-expression of the peptide core of gastric (or fetal colonic) mucins.

Flow Cytometry: 0,5-1 µg/million cells IF: 1 - 2 µg/ml IHC (FFPE): 0,5 - 1 µg/ml for 30 minutes at RT (1) Prediluted format: incubate for 30 min at RT (2) The concentration stated for each application is a general starting point. Variations in protocols, secondaries and substrates may require the MUC5AC antibody to be titered up or down for optimal performance.

1. Staining of formalin-fixed tissues requires boiling tissue sections in 10 mM citrate buffer, pH 6,0, for 10 - 20 min followed by cooling at RT for 20 minutes.
2. The prediluted format is supplied in a dropper bottle and is optimised for use in IHC. After epitope retrieval step (if required), drip mAb solution onto the tissue section and incubate at RT for 30 min.


Type: Primary
Antigen: MUC5AC
Clonality: Monoclonal
Clone: 45M1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human, Monkey, Rabbit, Cat, Mouse, Rat, Pig, Hedgehog, Chicken
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR