Anti-PGR Mouse Monoclonal Antibody [clone: PR484]
Lieferant: ProSci Inc.
33-188
PRSI33-188EA 773 EUR
PRSI33-188
Anti-PGR Mouse Monoclonal Antibody [clone: PR484]
Antikörper
This antibody is specific to progesterone receptor and shows minimal cross-reaction with other members of the family. Progesterone receptor is expressed as two major isoforms, PR-A (81 kDa) and PR-B (116 kDa). Expression of progesterone receptor has been suggested to reflect a intact estrogen regulatory machinery and therefore predict better clinical response to endocrine therapy than ER alone. This progesterone receptor antibody is excellent for immunohistochemical staining of formalin/paraffin tissues.
IHC (FFPE): 0,5 - 1 µg/ml for 30 min at RT (1) Prediluted format: incubate for 30 minMore Product Information
Neuer Artikel bei VWR!